Learn More
Netilmicin sulfate, MedChemExpress
MedChemExpress Netilmicin (sulfate) (SCH-20569 (sulfate)) is an active aminoglycoside antibiotic against most Gram-negative and some Gram-positive bacteria, including certain strains resistant to gentamicin.
Brand: Medchem Express HY-A0086
This item is not returnable.
View return policy
Description
Netilmicin (sulfate) (SCH-20569 (sulfate)) is an active aminoglycoside antibiotic against most Gram-negative and some Gram-positive bacteria, including certain strains resistant to gentamicin.Specifications
Netilmicin sulfate | |
Yellow | |
C21H41N5O7.5/2H2SO4 | |
SCH-20569 sulfate | |
NCC1=CC[C@@H](N)[C@](O[C@@H]2[C@@H](N)C[C@@H](NCC)[C@H](O[C@@]3([H])OC[C@](C)(O)[C@H](NC)[C@H]3O)[C@H]2O)([H])O1.[2.5H2SO4] | |
720.78 | |
Research | |
COVID-19-immunoregulation |
56391-57-2 | |
Research | |
5 mg | |
H2O : 100 mg/mL (138.74 mM; Need ultrasonic) ∣DMSO : 1 mg/mL (1.39 mM; ultrasonic and warming and heat to 80°C) | |
720.78 | |
98.0% | |
Solid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Research purposes only