Learn More
BRAND™ Natural Rubber Stoppers
Protects liquids from outside contaminants. Provides a secure seal.
Brand:  BRAND™ 144382
15.00 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.
 View more versions of this product
Product Code. 15271439
  • € 15.40 / Pack of 25
In Stock
to see stock.
Explore more special offers
Description
  • Natural rubber construction
  • Red color
  • Supplied in packs of 25
TRUSTED_SUSTAINABILITY
Specifications
Specifications
20 mm
Red
12 mm
25/PK
8 mm
Natural Rubber
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
BRAND™ Natural Rubber Stoppers > Upper dia.: 12mm; Base dia.: 8mm