missing translation for 'onlineSavingsMsg'
Learn More

BUCHI Glass Column MPLC

Brand:  BUCHI 022955

 View more versions of this product

Product Code. 10839770

This item is not returnable. View return policy

Description

Description

Specifications

Specifications

100 mm
C-690
Borosilicate Glass
460 mm
Sepacore™ C-690 MPLC Glass
MPLC Glass Column
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
BUCHI Glass Column MPLC > Length: 460mm; Dia.: 100mm

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.