Learn More
N-(4-Bromophenyl)benzenesulfonamide, 97%
Product Code. 15481738
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
Product Code. 15481738
Brand: Thermo Scientific Alfa Aesar H55476.03
Request Bulk or Custom Format
CAS: 16468-97-6 | C12H10BrNO2S | 312.181 g/mol
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 16468-97-6
Molecular Formula C12H10BrNO2S
Molecular Weight (g/mol) 312.181
MDL Number MFCD00159375
InChI Key HHLFFQAEABRCOJ-UHFFFAOYSA-N
Synonym n-4-bromophenyl benzenesulfonamide, benzenesulfonamide, n-4-bromophenyl, 4'-bromobenzenesulfonanilide, benzenesulfonanilide, 4'-bromo, 4-bromophenyl phenylsulfonyl amine, 4/'-bromobenzenesulfonanilide, n-p-bromophenylbenzenesulphonamide, n-4-bromophenyl benzenesulfonamide #
PubChem CID 293101
IUPAC Name N-(4-bromophenyl)benzenesulfonamide
SMILES C1=CC=C(C=C1)S(=O)(=O)NC2=CC=C(C=C2)Br
Specifications
Quantity 1 g
Formula Weight 312.18
Percent Purity 97%
Chemical Name or Material N-(4-Bromophenyl)benzenesulfonamide