Learn More
MP Biomedicals™ IPTG Powder
Induces of α-galactosidase activity in bacteria; suitable for use with X-gal or bluo-gal to detect lac gene activity in cloning procedure
€ 101.00 - € 402.00
Specifications
Content And Storage -20°C
pH 5 to 7
View More Specs
Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11347849
View Documents
MP Biomedicals™
114064112
5 g
€ 402.00
5g
11337849
View Documents
MP Biomedicals™
114064102
1 g
€ 101.00
1g
Description
Description
Protein trafficking; Cell viability; Cytotoxicity; Gene expression; Transformation; Cloning and expression
Specifications
Specifications
-20°C
5 to 7
Videos
Safety and Handling
Show More
SDS
Documents
Documents
Product Certifications
Product Content Correction
Product Title
MP Biomedicals™ IPTG Powder