Learn More
MP Biomedicals™ IPTG Powder
Induces of α-galactosidase activity in bacteria; suitable for use with X-gal or bluo-gal to detect lac gene activity in cloning procedure
Brand: MP Biomedicals™ 114064112
Description
Protein trafficking; Cell viability; Cytotoxicity; Gene expression; Transformation; Cloning and expression
Specifications
-20°C | |
5 to 7 | |
5 g |
Safety and Handling
- Isopropyl-beta-D-thiogalactopyranoside
Signal Word
- Warning
Hazard Category
- Carcinogenicity Category 2
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- H351-Suspected of causing cancer.
Precautionary Statement
- P102-Keep out of reach of children.
- P103-Read label before use.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P264-Wash thoroughly after handling.
- P272-Contaminated work clothing should not be allowed out of the workplace.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P321-Specific treatment (see on this label).
- P405-Store locked up.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
- MIXTURE LIST-Contains : Isopropyl-lactopyranoside