Learn More
SCHOTT AG Lighting and Imaging™ Mounting Bracket for EasyLED Spotlight Plus and KL300LED
Shop All Schott Products
Click to view available options
For Use With (Equipment):
25 mm dia. Column
30 mm dia. Column
32 mm dia. Column
35 mm dia. Column
Base 158.340
Zeiss Stand K, Stand M
Unit Size:
1 unit
Each
Description
- Connects the EasyLED Spotlight Plus or KL300LED to the microscope column, base or a base.
Specifications
Specifications
| Type | Zeiss Stand K and M |
| For Use With (Application) | Stereo Microscopy Illumination |
| For Use With (Equipment) | Zeiss Stand K, Stand M |