missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Monomethyl fumarate, Thermo Scientific™
€ 48.30 - € 152.00
Specifications
Chemical Name or Material | Monomethyl fumarate |
---|---|
CAS | 2756-87-8 |
Melting Point | 145°C to 150°C |
Shipping Condition | No special requirements |
Product Line | Building blocks |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
17760669
|
Thermo Scientific Acros
469750050 |
5 g |
€ 48.30
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
17770669
|
Thermo Scientific Acros
469750250 |
25 g |
€ 152.00
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Specifications
Monomethyl fumarate | |
145°C to 150°C | |
Building blocks | |
Powder or Crystals | |
≥96.0% |
2756-87-8 | |
No special requirements | |
C5 H6 O4 | |
White to Almost-white |
Safety and Handling
Exclamation mark
Corrosion
missing translation for 'einecsNumber' : 220-412-6
Recommended Storage : Normal conditions