missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Monomethyl fumarate, Thermo Scientific™
Brand: Thermo Scientific Acros 469750250
This item is not returnable.
View return policy
Description
Specifications
Monomethyl fumarate | |
25 g | |
No special requirements | |
C5 H6 O4 | |
White to Almost-white |
2756-87-8 | |
145°C to 150°C | |
Building blocks | |
Powder or Crystals | |
≥96.0% |