Learn More
ML171, MedChemExpress
Product Code. 16465986
Encompass_Preferred
Click to view available options
Quantity:
100 mg
500 mg
Unit Size:
100mg
500mg
Product Code. 16465986
Brand: Medchem Express HY12805
MedChemExpress ML171 (2-Acetylphenothiazine;2-APT) is a potent and selective NADPH oxidase 1 (Nox1) inhibitor that blocks Nox1-dependent ROS generation, with an IC50 of 0.25 μM in HEK293-Nox1 confirmatory assay.
ML171 (2-Acetylphenothiazine
Specifications
Chemical Name or Material ML171
CAS 6631-94-3
Color Orange
Purity Grade Notes Research
Molecular Formula C14H11NOS
Quantity 100 mg
Synonym 2-Acetylphenothiazine 2-APT
Solubility Information DMSO : ≥ 64 mg/mL (265.22 mM)
SMILES CC(C1=CC=C2SC3=CC=CC=C3NC2=C1)=O
Molecular Weight (g/mol) 241.31
Formula Weight 241.31
Percent Purity 99.9%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation
Show More Show Less
Research purposes only