Learn More
Greiner Bio-One Microlon™ Polystyrene 96-well ELISA Microplates
Manufactured from crystal clear polystyrene for optimum visibility. Greiner BioOne™ Microlon™ Polystyrene 96-Well ELISA Microplates are tested in a colorimetric immunoasay for immunological research applications.
€ 66.29 - € 85.81
Specifications
Format | Standard |
---|---|
Lid | Without Lid |
Array | 8 x 12 |
Color | Clear |
Material | Polystyrene |
Product Code | Brand | Binding Property | Surface Treatment | Well Design | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Binding Property | Surface Treatment | Well Design | Price | Quantity & Availability | ||||
10577091
|
Greiner Bio-One
655061 |
High Binding | Microlon 600 | Standard Well |
€ 85.81
Pack of 40 |
Estimated Shipment: 07-05-2025
Log in to see stock. |
||||
10144771
|
Greiner Bio-One
655001 |
Medium Binding | Microlon 200 | Standard Well |
€ 66.29
Pack of 40 |
Estimated Shipment: 13-05-2025
Log in to see stock. |
||||
10680932
|
Greiner Bio-One
655080 |
Medium Binding | Microlon 200 | Chimney Well |
€ 85.81
Pack of 40 |
Estimated Shipment: 15-05-2025
Log in to see stock. |
||||
Description
- Free of detectable DNase, RNase, human DNA
- Non-Pyrogenic
- Manufactured under DIN ISO 9001 guidelines
- Tracebility from production through a defined LOT number system
- Manufactured from crystal clear polystyrene
- The footprint is compatible with automated systems
- Well to well spacing : 9 mm
- Alphanumeric well coding
- Suitable for transmission measurements
- Specifically treated to provide an incresed protein binding
- Well profile: F-bottom / standard (flat bottom)
Specifications
Standard | |
8 x 12 | |
Polystyrene | |
Flat Bottom (F-well) |
Without Lid | |
Clear | |
96 | |
40/Cs. |
Product Title
Greiner Bio-One Microlon™ Polystyrene 96-well ELISA Microplates