Learn More
Greiner Bio-One Microlon™ Polystyrene 96-well ELISA Microplates
Click to view available options
Binding Property:
High Binding
Medium Binding
Surface Treatment:
Microlon 200
Microlon 600
Well Design:
Chimney Well
Standard Well
Conditionnement:
Pack of 40
Description
- Free of detectable DNase, RNase, human DNA
- Non-Pyrogenic
- Manufactured under DIN ISO 9001 guidelines
- Tracebility from production through a defined LOT number system
- Manufactured from crystal clear polystyrene
- The footprint is compatible with automated systems
- Well to well spacing : 9 mm
- Alphanumeric well coding
- Suitable for transmission measurements
- Specifically treated to provide an incresed protein binding
- Well profile: F-bottom / standard (flat bottom)
Spécification
Spécification
| Binding Property | Medium Binding |
| Format | Standard |
| Lid | Without Lid |
| Array | 8 x 12 |
| Color | Clear |
| Material | Polystyrene |
| No. of Wells | 96 |
| Sterility | Non-sterile |
| Surface Treatment | Microlon 200 |
| Well Shape | Flat Bottom (F-well) |
| Afficher plus |