Learn More
Methyl 3-amino-4-methylbenzoate, 97%, Thermo Scientific Chemicals
€ 18.50 - € 59.00
Chemical Identifiers
CAS | 18595-18-1 |
---|---|
Molecular Formula | C9H11NO2 |
Molecular Weight (g/mol) | 165.19 |
MDL Number | MFCD00025206 |
InChI Key | YEPWCJHMSVABPQ-UHFFFAOYSA-N |
Synonym | 3-amino-4-methylbenzoic acid methyl ester, methyl 3-amino-4-methyl-benzoate, benzoic acid, 3-amino-4-methyl-, methyl ester, methyl 3-amino-p-toluate, methyl 4-methyl-3-aminobenzoate, 3-amino-p-toluic acid methyl ester, 3-amino-4-methyl-benzoic acid methyl ester, 3-amino-4-methyl benzoic acid methyl ester, pubchem2351, pubchem10930Show More |
PubChem CID | 337778 |
IUPAC Name | methyl 3-amino-4-methylbenzoate |
SMILES | COC(=O)C1=CC=C(C)C(N)=C1 |
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
18595-18-1 | |
165.19 | |
YEPWCJHMSVABPQ-UHFFFAOYSA-N | |
337778 | |
COC(=O)C1=CC=C(C)C(N)=C1 |
C9H11NO2 | |
MFCD00025206 | |
3-amino-4-methylbenzoic acid methyl ester, methyl 3-amino-4-methyl-benzoate, benzoic acid, 3-amino-4-methyl-, methyl ester, methyl 3-amino-p-toluate, methyl 4-methyl-3-aminobenzoate, 3-amino-p-toluic acid methyl ester, 3-amino-4-methyl-benzoic acid methyl ester, 3-amino-4-methyl benzoic acid methyl ester, pubchem2351, pubchem10930Show More | |
methyl 3-amino-4-methylbenzoate |
Specifications
18595-18-1 | |
White to Cream | |
96% min. (HPLC) | |
CH3(NH2)C6H3COOCH3 | |
3-amino-4-methylbenzoic acid methyl ester, methyl 3-amino-4-methyl-benzoate, benzoic acid, 3-amino-4-methyl-, methyl ester, methyl 3-amino-p-toluate, methyl 4-methyl-3-aminobenzoate, 3-amino-p-toluic acid methyl ester, 3-amino-4-methyl-benzoic acid methyl ester, 3-amino-4-methyl benzoic acid methyl ester, pubchem2351, pubchem10930 | |
COC(=O)C1=CC=C(C)C(N)=C1 | |
165.19 | |
165.19 | |
Crystalline Powder |
114.0°C to 117.0°C | |
Authentic | |
C9H11NO2 | |
MFCD00025206 | |
YEPWCJHMSVABPQ-UHFFFAOYSA-N | |
methyl 3-amino-4-methylbenzoate | |
337778 | |
97% | |
Methyl 3-amino-4-methylbenzoate |
Safety and Handling
GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for
GHS Signal Word: Warning
- Methyl 3-amino-4-methylbenzoate
Signal Word
- Warning
Hazard Category
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- H315-Causes skin irritation.
Precautionary Statement
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.