missing translation for 'onlineSavingsMsg'
Learn More
Methyl 3-amino-4-methylbenzoate, 97%, Thermo Scientific Chemicals Product Code.: 11468266

Methyl 3-amino-4-methylbenzoate, 97%, Thermo Scientific Chemicals

Product Code. 11468266
25 g
Click to view available options
Quantity:
5 g
25 g
100 g
This item is not returnable. View return policy

Product Code. 11468266

Brand: Thermo Scientific Alfa Aesar A15348.14

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 18595-18-1
Molecular Formula C9H11NO2
Molecular Weight (g/mol) 165.19
MDL Number MFCD00025206
InChI Key YEPWCJHMSVABPQ-UHFFFAOYSA-N
Synonym 3-amino-4-methylbenzoic acid methyl ester, methyl 3-amino-4-methyl-benzoate, benzoic acid, 3-amino-4-methyl-, methyl ester, methyl 3-amino-p-toluate, methyl 4-methyl-3-aminobenzoate, 3-amino-p-toluic acid methyl ester, 3-amino-4-methyl-benzoic acid methyl ester, 3-amino-4-methyl benzoic acid methyl ester, pubchem2351, pubchem10930Show MoreShow Less
PubChem CID 337778
IUPAC Name methyl 3-amino-4-methylbenzoate
SMILES COC(=O)C1=CC=C(C)C(N)=C1

Specifications

Melting Point 113°C to 117°C
Quantity 25 g
Beilstein 2088611
Formula Weight 165.19
Percent Purity 97%
Chemical Name or Material Methyl 3-amino-4-methylbenzoate

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number5004P27S Certificate TypeCertificate of Analysis Date23/01/2024 Catalog NumberA15348.06,A15348.14,A15348.22
Lot NumberJ6392A Certificate TypeCertificate of Analysis Date22/11/2021 Catalog NumberA15348.06,A15348.14,A15348.22
Lot Number5004P27S Certificate TypeCertificate of Analysis Date22/11/2021 Catalog NumberA15348.06,A15348.14,A15348.22
Lot Number5002L24X Certificate TypeCertificate of Analysis Date22/11/2021 Catalog NumberA15348.06,A15348.14,A15348.22
Lot NumberJ3309A Certificate TypeCertificate of Analysis Date22/11/2021 Catalog NumberA15348.06,A15348.14,A15348.22

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Methyl 3-amino-4-methylbenzoate, 97%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.