missing translation for 'onlineSavingsMsg'
Learn More
(Methoxycarbonylmethyl)triphenylphosphonium bromide, 98+%, Thermo Scientific Chemicals Product Code.: 11467767

(Methoxycarbonylmethyl)triphenylphosphonium bromide, 98+%, Thermo Scientific Chemicals

Product Code. 11467767
100 g
Click to view available options
Quantity:
25 g
100 g
500 g
This item is not returnable. View return policy

Product Code. 11467767

Brand: Thermo Scientific Alfa Aesar A16301.22

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

(Methoxycarbonylmethyl)triphenylphosphonium bromide is a reactant for the synthesis of GSK-3 inhibitors using the Wittig reaction, substituted furanones for antifungal and cytostatic activities, photochromic dithienylethene derivatives, gem-diiodoalkanes containing allylic alcohols and highly substituted cyclopentenes and brazilanes via Au-catalyzed deoxygenative Nazarov cyclization. As reactant for asymmetric hydrogenation of α,β-unsaturated ester-phosphonates, studies of thermal decomposition of phosphonium alkyl ester salts.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
(Methoxycarbonylmethyl)triphenylphosphonium bromide is a reactant for the synthesis of GSK-3 inhibitors using the Wittig reaction, substituted furanones for antifungal and cytostatic activities, photochromic dithienylethene derivatives, gem-diiodoalkanes containing allylic alcohols and highly substituted cyclopentenes and brazilanes via Au-catalyzed deoxygenative Nazarov cyclization. As reactant for asymmetric hydrogenation of α,β-unsaturated ester-phosphonates, studies of thermal decomposition of phosphonium alkyl ester salts.

Solubility
Soluble in methanol(almost transparency).

Notes
Hygroscopic. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, water, moisture.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1779-58-4
Molecular Formula C21H20BrO2P
Molecular Weight (g/mol) 415.27
MDL Number MFCD00011801
InChI Key VCWBQLMDSMSVRL-UHFFFAOYSA-M
Synonym 2-methoxy-2-oxoethyl triphenylphosphonium bromide, methoxycarbonylmethyl triphenylphosphonium bromide, carbomethoxymethyl triphenylphosphonium bromide, methoxycarbonylmethyl triphenyl phosphonium bromide, 2-methoxy-2-oxoethyl triphenylphosphanium bromide, 2-methoxy-2-oxoethyl-triphenylphosphanium bromide, 2-methoxy-2-oxo-ethyl-triphenyl-phosphonium bromide, phosphonium, 2-methoxy-2-oxoethyl triphenyl-, bromide, acmc-209edxShow MoreShow Less
PubChem CID 2733211
IUPAC Name (2-methoxy-2-oxoethyl)-triphenylphosphanium;bromide
SMILES [Br-].COC(=O)C[P+](C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Melting Point ∼163°C (decomposition)
Quantity 100 g
Beilstein 3812975
Sensitivity Hygroscopic
Solubility Information Soluble in methanol(almost transparency).
Formula Weight 415.28
Percent Purity ≥98%
Chemical Name or Material (Methoxycarbonylmethyl)triphenylphosphonium bromide

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

4 results found
Lot Number Certificate Type Date Catalog Number
Lot Number10236940 Certificate TypeCertificate of Analysis Date30/08/2024 Catalog NumberA16301.22,A16301.14,A16301.36
Lot Number10137317 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberA16301.22,A16301.14,A16301.36
Lot Number10160936 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberA16301.22,A16301.14,A16301.36
Lot Number10236940 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberA16301.22,A16301.14,A16301.36

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
(Methoxycarbonylmethyl)triphenylphosphonium bromide, 98+%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.