Learn More
Magnesium tert-butoxide, 90+%, Thermo Scientific Chemicals
€ 37.00 - € 353.76
Chemical Identifiers
CAS | 32149-57-8 |
---|---|
Molecular Formula | C8H18MgO2 |
Molecular Weight (g/mol) | 170.535 |
MDL Number | MFCD03788761 |
InChI Key | RTKCPZYOLXPARI-UHFFFAOYSA-N |
Synonym | magnesium tert-butoxide, magnesium di-tert-butoxide, magnesium 2-methylpropan-2-olate, magnesium t-butoxide, magnesiumtert-butoxide, di-tert-butoxymagnesium, magnesium tert. butoxide, ksc226k7n, magnesium 2+ bis 2-methylpropan-2-olateShow More |
PubChem CID | 12040442 |
IUPAC Name | magnesium;2-methylpropan-2-olate |
SMILES | CC(C)(C)[O-].CC(C)(C)[O-].[Mg+2] |
Product Code | Brand | Quantity | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | ||||||
11345615
|
Thermo Scientific Alfa Aesar
L19745.09 |
10 g | N/A | N/A | N/A | |||||
11359675
|
Thermo Scientific Alfa Aesar
L19745.18 |
50 g | N/A | N/A | N/A | |||||
11369675
|
Thermo Scientific Alfa Aesar
L19745.30 |
250 g |
€ 353.76
250g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||||
Description
Magnesium tert-butoxide is used as a pharmaceutical intermediate. It is also used in deprotonation reactions for organic syntheses.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsMagnesium tert-butoxide is used as a pharmaceutical intermediate. It is also used in deprotonation reactions for organic syntheses.
Solubility
Reacts with water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with bases and oxidizing agents. Store under dry inert gas. It is sensitive to air and moisture.
Chemical Identifiers
32149-57-8 | |
170.535 | |
RTKCPZYOLXPARI-UHFFFAOYSA-N | |
12040442 | |
CC(C)(C)[O-].CC(C)(C)[O-].[Mg+2] |
C8H18MgO2 | |
MFCD03788761 | |
magnesium tert-butoxide, magnesium di-tert-butoxide, magnesium 2-methylpropan-2-olate, magnesium t-butoxide, magnesiumtert-butoxide, di-tert-butoxymagnesium, magnesium tert. butoxide, ksc226k7n, magnesium 2+ bis 2-methylpropan-2-olateShow More | |
magnesium;2-methylpropan-2-olate |
Specifications
32149-57-8 | |
[(CH3)3CO]2Mg | |
UN2921 | |
Air and moisture sensitive | |
Reacts with water. | |
CC(C)(C)[O-].CC(C)(C)[O-].[Mg+2] | |
170.535 | |
170.54 | |
Magnesium tert-butoxide |
C8H18MgO2 | |
MFCD03788761 | |
3979348 | |
magnesium tert-butoxide, magnesium di-tert-butoxide, magnesium 2-methylpropan-2-olate, magnesium t-butoxide, magnesiumtert-butoxide, di-tert-butoxymagnesium, magnesium tert. butoxide, ksc226k7n, magnesium 2+ bis 2-methylpropan-2-olate | |
RTKCPZYOLXPARI-UHFFFAOYSA-N | |
magnesium;2-methylpropan-2-olate | |
12040442 | |
≥90% |
Safety and Handling
GHS H Statement
H314-H228
Causes severe skin burns and eye damage.
Flammable solid.
P210-P240-P241-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P370+P378q-P501c
H228-H315-H319-H332-H335
missing translation for 'dotInformation' : Hazard Class: 4.2; Packaging Group: II
missing translation for 'einecsNumber' : 250-931-3
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.