missing translation for 'onlineSavingsMsg'
Learn More

Magnesium tert-butoxide, 90+%

Product Code. 11359675
Click to view available options
Quantity:
10 g
50 g
250 g
Unit Size:
10g
250g
50g
This item is not returnable. View return policy
Regulatory: This product is an isolated intermediate transported in accordance with Regulation (EC) 1907/2006 (REACH). All customers must complete a declaration before delivery.

Product Code. 11359675

Brand: Thermo Scientific Alfa Aesar L19745.18

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy
Regulatory: This product is an isolated intermediate transported in accordance with Regulation (EC) 1907/2006 (REACH). All customers must complete a declaration before delivery.

CAS: 32149-57-8 | C8H18MgO2 | 170.535 g/mol

Magnesium tert-butoxide is used as a pharmaceutical intermediate. It is also used in deprotonation reactions for organic syntheses.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Magnesium tert-butoxide is used as a pharmaceutical intermediate. It is also used in deprotonation reactions for organic syntheses.

Solubility
Reacts with water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with bases and oxidizing agents. Store under dry inert gas. It is sensitive to air and moisture.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 32149-57-8
Molecular Formula C8H18MgO2
Molecular Weight (g/mol) 170.535
MDL Number MFCD03788761
InChI Key RTKCPZYOLXPARI-UHFFFAOYSA-N
Synonym magnesium tert-butoxide, magnesium di-tert-butoxide, magnesium 2-methylpropan-2-olate, magnesium t-butoxide, magnesiumtert-butoxide, di-tert-butoxymagnesium, magnesium tert. butoxide, ksc226k7n, magnesium 2+ bis 2-methylpropan-2-olate
PubChem CID 12040442
IUPAC Name magnesium;2-methylpropan-2-olate
SMILES CC(C)(C)[O-].CC(C)(C)[O-].[Mg+2]

Specifications

Linear Formula [(CH3)3CO]2Mg
Quantity 50 g
UN Number UN2921
Beilstein 3979348
Sensitivity Air and moisture sensitive
Solubility Information Reacts with water.
Formula Weight 170.54
Percent Purity ≥90%
Chemical Name or Material Magnesium tert-butoxide

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.