Learn More
LW6, MedChemExpress
MedChemExpress LW6 (HIF-1α inhibitor) is a novel HIF-1 inhibitor with an IC50 of 4.4 μM. LW6 decreases HIF-1α protein expression without affecting HIF-1β expression.
Brand: Medchem Express HY-13671
This item is not returnable.
View return policy
Description
LW6 (HIF-1α inhibitor) is a novel HIF-1 inhibitor with an IC50 of 4.4 μM. LW6 decreases HIF-1α protein expression without affecting HIF-1β expression.Specifications
LW6 | |
Off-White | |
C26H29NO5 | |
HIF-1α inhibitor LW8 | |
O=C(OC)C1=CC=C(O)C(NC(COC2=CC=C(C3(C4)CC5CC4CC(C5)C3)C=C2)=O)=C1 | |
435.51 | |
Research | |
Cancer-programmed cell death |
934593-90-5 | |
Research | |
50 mg | |
DMSO : 25 mg/mL (57.40 mM; Need ultrasonic) ∣DMF : 17.24 mg/mL (39.59 mM; Need ultrasonic) | |
435.51 | |
98.0% | |
Solid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Research purposes only