Learn More
Fisherbrand™ Lab Thermometer
A general purpose/stirring thermometer for a wide application of laboratory needs.
Brand:  Fisherbrand™ FB58553
 View more versions of this product
Product Code. 10121901
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Explore more special offers
Description
Description
  • With amber graduations
  • Non-roll fitting
  • Reinforced bulb for stirring
Specifications
Specifications
General use
155 mm
Liquid Filled Thermometer
-10°C
1°C
Total Immersion
Red Spirit
50°C
-10°C to +50°C