Learn More
Fisherbrand™ Lab Thermometer
A general purpose/stirring thermometer for a wide application of laboratory needs.
€ 3.15 - € 7.70
Specifications
For Use With (Application) | General use |
---|---|
Thermometer Fill | Red Spirit |
Product Type | Liquid Filled Thermometer |
Product Code | Brand | Temperature Range (Metric) | Subdivision | Immersion Type | Length (Metric) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Temperature Range (Metric) | Subdivision | Immersion Type | Length (Metric) | Price | Quantity & Availability | |||||
10672272
|
Fisherbrand™
FB58543 |
-20°C to +110°C | 1°C | Total Immersion | 305 mm | N/A | ||||||
11587273
|
Fisherbrand™
63/019/1 |
-20°C to +110°C | 1°C | Total Immersion | 305 mm |
€ 4.11
Each |
In Stock
Log in to see stock. ![]() |
|||||
10498202
|
Fisherbrand™
FB58545 |
-20°C to +150°C | 1°C | Total Immersion | 305 mm | N/A | ||||||
11597273
|
Fisherbrand™
63/020/1 |
-20°C to +150°C | 1°C | Total Immersion | 305 mm |
€ 4.84
Each |
In Stock
Log in to see stock. ![]() |
|||||
10121901
|
Fisherbrand™
FB58553 |
-10°C to +50°C | 1°C | Total Immersion | 155 mm | N/A | ||||||
10559781
|
Fisherbrand™
FB58555 |
-10°C to +110°C | 1°C | Total Immersion | 155 mm | N/A | ||||||
11517283
|
Fisherbrand™
63/022/1 |
-10°C to +110°C | 1°C | Total Immersion | 155 mm |
€ 4.21
Each |
In Stock
Log in to see stock. ![]() |
|||||
11551993
|
Fisherbrand™
63/098/1 |
-10°C to +50°C | 0.5°C | Partial Immersion | 305 mm |
€ 4.81
Each |
In Stock
Log in to see stock. |
|||||
11561993
|
Fisherbrand™
63/098/2 |
-10°C to +110°C | 1°C | Partial Immersion | 305 mm |
€ 3.85
Each |
In Stock
Log in to see stock. ![]() |
|||||
11571993
|
Fisherbrand™
63/098/3 |
-10°C to +150°C | 1°C | Partial Immersion | 305 mm |
€ 4.89
Each |
In Stock
Log in to see stock. ![]() |
|||||
Description
- With amber graduations
- Non-roll fitting
- Reinforced bulb for stirring
Specifications
General use | |
Red Spirit | |
Liquid Filled Thermometer |
Product Title
Fisherbrand™ Lab Thermometer