Learn More
Kartell™ Latex Gas Sampling Balloon
Gass collection bags that are constructed from latex. Kartell™ Latex Gas Sampling Balloon are best suited for collecting, containing, and carrying gas samples.
Brand:  Kartell™ 0123500
 View more versions of this product
Product Code. 10151183
  • € 49.95 / Each
Estimated Shipment: 29-08-2025
to see stock.
Explore more special offers
Description
  • Cost effective
  • Ideal for collecting, containing, and carrying gas samples
Specifications
Specifications
Gas Sampling Balloon
8 L
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Kartell™ Latex Gas Sampling Balloon > Capacity: 8 L