Learn More
Kartell™ Latex Gas Sampling Balloon
Product Code. p-8007243
Click to view available options
Capacity (Metric):
1 L
1.5 L
2 L
4 L
5 L
8 L
Unit Size:
Each
Product Code. 10680374
Brand: Kartell™ 0123000
Only null left
Add to basket
Add to basket
Gass collection bags that are constructed from latex. Kartell™ Latex Gas Sampling Balloon are best suited for collecting, containing, and carrying gas samples.
  • Cost effective
  • Ideal for collecting, containing, and carrying gas samples
Specifications
Capacity (Metric) 1 L
Material Latex
Diameter (Metric) 130 mm
Product Type Gas Sampling Balloon