Learn More
Buerkle™ LaboPlast™ Bio Disposable Scoop
BIOPHARMAGREENER_CHOICE
Produced in a Cleanroom Class 7 (10,000) and individually wrapped in bio-PE film for disposable use. Green PE bio-plastic has all the suitable properties of conventional PE but is manufactured from renewable raw materials.
Brand:  Buerkle™ 5379-0005
 View more versions of this product
Product Code. 15386327
  • € 28.20 / Pack of 10
In Stock
to see stock.
Explore more special offers
Description
  • Green PE bio-plastic has all the suitable properties of conventional polyethylene but is manufactured from renewable raw materials and can be completely recycled
  • The scoop stands horizontally as a result of the special handle design and the slightly angled front edge and can therefore be put down without the sample material trickling out.
  • Product and packaging made of renewable raw materials
  • Bio-PE, white
  • Individually wrapped
  • Production and packaging according to Cleanroom Class 7
  • With EU foodstuffs and FDA approval
Specifications
Specifications
Polyethylene
Single-use
White
Foodstuffs
205 mm
100 mL
Non-sterile
FDA
10
Videos
  • Buerkle LaboPlast Steriplast Bio Disposable Scoop
    Buerkle LaboPlast Steriplast Bio Disposable Scoop
    Video Player is loading.
    Current Time 0:00
    Duration 0:00
    Loaded: 0%
    Stream Type LIVE
    Remaining Time 0:00
    1x
    • Chapters
    • descriptions off, selected
    • captions off, selected
      Buerkle LaboPlast Steriplast Bio Disposable Scoop
    SDS
    Documents Certificates
    Certificates
    A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
    5 results found
    Lot Number Certificate Type Date Catalog Number
    Lot Number251402 Certificate TypeCertificate of Compliance Date11/04/2025 Catalog Number5379-0005
    Lot Number243293 Certificate TypeCertificate of Compliance Date16/08/2024 Catalog Number5379-0005
    Lot Number240666 Certificate TypeCertificate of Compliance Date21/02/2024 Catalog Number5379-0005
    Lot Number233913 Certificate TypeCertificate of Compliance Date05/09/2023 Catalog Number5379-0005
    Lot Number230767 Certificate TypeCertificate of Compliance Date23/02/2023 Catalog Number5379-0005
    Special Offers
    Special Offers
    Product Content Correction
    Product Title
    Buerkle™ LaboPlast™ Bio Disposable Scoop > 100mL