Learn More
DWK Life Sciences Wheaton™ Dounce Tissue Grinders
Ideal for enzyme studies. Wheaton™ Dounce Tissue Grinders are designed to retain a high percentage of cell nuclei, mitochondria, and proteins in soft tissues or cell suspensions.
Brand: DWK Life Sciences 357546
Description
Ideal for enzyme studies. Wheaton™ Dounce Tissue Grinders are designed to retain a high percentage of cell nuclei, mitochondria, and proteins in soft tissues or cell suspensions.
- Sample covers the pestle ball, preventing heat buildup by reducing friction against the mortar
- Mortar features large reservoir and pouring lip
- Supplied with “loose” and “tight” pestles
- Components are made of low extractable borosilicate glass
Order Info
- Tight pestle clearance: 0.0020 ±0.001 in. (0.05 ±0.025mm)
- Loose pestle clearance: 0.114 ±0.001 in. (0.114 ±0.025mm)

Specifications
1.35 oz. | |
Borosilicate Glass | |
Designed to retain a high percentage of cell nuclei, mitochondria and Proteins in soft tissues or cell suspensions | |
285mm | |
140mm | |
32mm |
40 mL | |
Dounce Tissue Grinder | |
11.2 in. | |
5.5 in. | |
1.25 in. | |
1.35 oz., 40mL |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.