Learn More
Kontes™ Membrane Filtration Glassware from Restek
Product Code. 11888512
Click to view available options
Type:
Flask (2 L)
Flask (4 L)
Funnel (1 L)
Funnel (300 mL)
Funnel (500 mL)
Support
Material:
Fritted Glass
Glass
Unit Size:
Each
Product Code. 11888512
Brand: Kimble Chase 9538280000
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Used for routine filtration of corrosive liquids and removal of particles from HPLC solvents
  • Ground joint connection eliminates phthalate contamination that can occur when using silicone or neoprene stoppers
  • Support base has a coarse porosity glass frit and an integral vacuum connection, located above the drip tip that prevents contamination of the vacuum line with filtrate droplets
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Equipment) Microfiltration Apparatus
Material Glass
Type Flask (2 L)
Volume (Metric) 2 L