missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
Inactive analog of the CaM kinase II inhibitor KN 93 . Potassium channel (Kv1.2, 1.4, 1.5, 2.1, 3.2 and hERG) blocker in vitro.
Active Analog also available.

Specifications
Specifications
CAS | 1135280-28-2 |
Molecular Formula | C24H28ClN2O7PS |
Quantity | 1 mg |
Synonym | kn-92 phosphate, kn92-h3po4, 2-n-4-methoxybenzenesulfonyl amino-n-4-chlorocinnamyl-n-methylbenzylamine, phosphate, kn92 phosphate, c24h25cln2o3s.h3o4p, kn 92 phosphate;kn92 phosphate, phosphate, 2-n-4'-methoxybenzenesulfonyl amino-n-4'-chlorophenyl-2-propenyl-n-methylbenzylamine phosphate, phosphoric acid-n-2-2e-3-4-chlorophenyl prop-2-en-1-yl methyl amino methyl phenyl-4-methoxybenzene-1-sulfonamide 1/1 |
InChI Key | XRQHWVVDNMJDEQ-IPZCTEOASA-N |
SMILES | CN(CC=CC1=CC=C(C=C1)Cl)CC2=CC=CC=C2NS(=O)(=O)C3=CC=C(C=C3)OC.OP(=O)(O)O |
IUPAC Name | N-[2-[[[(E)-3-(4-chlorophenyl)prop-2-enyl]-methylamino]methyl]phenyl]-4-methoxybenzenesulfonamide;phosphoric acid |
Molecular Weight (g/mol) | 554.979 |
PubChem CID | 16219540 |
Formula Weight | 554.98 |
Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
KN 92, Tocris Bioscience™ >
Spot an opportunity for improvement?Share a Content Correction