Learn More
Thermo Scientific™ Kits and Accessories for Orion™ 2117XP Chloride Analyzers
Keep chloride analyzers well-supplied and ready for use with the Thermo Scientific™ Kits and Accessories for Orion™ 2117XP Chloride Analyzers.
Brand: Thermo Scientific™ 2117XPEK
View more versions of this product
Description
Keep chloride analyzers well-supplied and ready for use with the Thermo Scientific™ Kits and Accessories for Orion™ 2117XP Chloride Analyzers. Choose from the reagent-less consumables kit, reagent bottle adapter, chloride calibration kit or electrode kit.
Warranty: Out of box
Specifications
Chloride Electrode Kit | |
Orion™ 2117XP Chloride Analyzers |
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.