Learn More
Brady™ BMP71 Label Printer Accessory: Cleaning Kit
Cleaning Kit for use with the BMP Label Printer.
Brand: Brady™ 114886
Description
- For use in cleaning the BMP™71 Label Printer (sold separately)
- Includes printer cleaning cards and swabs
- Printer cleaning kits provide an easy solution for keeping your printer in top condition
- Provides a safe and easy way to clean your printer
Specifications
Cleaning Kit | |
M71-CLEAN | |
Cleaning Kit | |
1 Ea. |
Safety and Handling
- EZ Clean 01 (99.7%) Free Flowing and Non-Free Flowing
Signal Word
- Danger
Hazard Category
- Serious eye damage/eye irritation Category 2
- Flammable liquid Category 2
Hazard Statement
- H225-Highly flammable liquid and vapour.
- H318-Causes serious eye damage.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P233-Keep container tightly closed.
- P264-Wash thoroughly after handling.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P337+P313-If eye irritation persists: Get medical advice/attention.
- P370+P378-In case of fire: Use water spray or fog,foam, carbon dioxideor dry chemical to extinguish.
- P403+P235-Store in a well-ventilated place. Keep cool.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
- MIXTURE LIST-Contains : isopropanol