Learn More
DWK Life Sciences Kimble™ Kontes™ Brand Microflex™ Microscale Kits - Syringe Needles
Nonsterile
Brand: DWK Life Sciences 868280-2201
This item is not returnable.
View return policy
Description
- Size: 1 in.
- Sold 12/pk
Specifications
Stainless Steel | |
0.028 in. | |
For general purpose dispensing in laboratory applications and for microflex microscale kits | |
Syringe Needle |
22 G | |
0.7 mm | |
With an inert plastic luer hub and a blunt end for safety | |
Sterile |
Product Content Correction
Product Title
DWK Life Sciences Kimble™ Kontes™ Brand Microflex™ Microscale Kits - Syringe Needles > Syringe Needle, Gauge: 22