Learn More
KG-501, MedChemExpress
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
50mg
5mg
Description
KG-501 is a CREB inhibitor, with an IC50 of 6.89 μM.
Specifications
Specifications
| Chemical Name or Material | KG-501 |
| CAS | 18228-17-6 |
| Color | Light Yellow |
| Purity Grade Notes | Research |
| Molecular Formula | C17H13ClNO5P |
| Quantity | 5 mg |
| Synonym | Naphthol AS-E phosphate |
| Solubility Information | DMSO : 6 mg/mL (15.88 mM; Need ultrasonic and warming) |
| SMILES | O=C(C1=C(OP(O)(O)=O)C=C2C=CC=CC2=C1)NC3=CC=C(Cl)C=C3 |
| Molecular Weight (g/mol) | 377.72 |
| Show More |
Research purposes only