Learn More
B Braun™ FEP Safety IV Catheters, Introcan
Use B Braun™ FEP Safety IV Catheters, Introcan™. From insertion to catheter deployment to needle removal, protection is provided by an automatically activated system that cannot be bypassed.
€ 88.00 - € 157.00
Specifications
Product Type Introcan
View More Specs
Products 3
Product Code Brand Needle Gauge Price Quantity & Availability  
Product Code Brand Needle Gauge Price Quantity & Availability  
12301113
View Documents
B Braun™
4254562-01
18 G
€ 157.00
Pack of 50
10453202
View Documents
B Braun™
4252071B
24 G
€ 89.00
Pack of 50
10174713
View Documents
B Braun™
4252098B
22 G
€ 88.00
Pack of 50
Description
Description
  • Color-coded labels for easy gauge size identification
  • Back cut bevel
  • FEP for firmer construction for arterial access and special procedures
  • DEHP- and latex-free
Specifications
Specifications
Introcan