Learn More
B Braun™ FEP Safety IV Catheters, Introcan
Use B Braun™ FEP Safety IV Catheters, Introcan™. From insertion to catheter deployment to needle removal, protection is provided by an automatically activated system that cannot be bypassed.
Brand: B Braun™ 4254562-01
This item is not returnable.
View return policy
Description
- Color-coded labels for easy gauge size identification
- Back cut bevel
- FEP for firmer construction for arterial access and special procedures
- DEHP- and latex-free
Specifications
18 G | |
32 mm | |
Introcan |