Learn More
Thermo Scientific™ Indiko™ and Konelab™ System Calibrators and Controls
Ensure accurate test results with Thermo Scientific™ Indiko™ and Konelab™ System Calibrators and Controls.
€ 229.00
Specifications
Certifications/Compliance Compliant with the IVDD; CE marked
Sample Type Serum, Urine
View More Specs
Products 2
Product Code Brand Description Price Quantity & Availability  
Product Code Brand Description Price Quantity & Availability  
10262285
View Documents
Thermo Scientific™
981821
uTrol Control
€ 229.00
Pack of 5
Estimated Shipment: 18-02-2025
to see stock.
Add to basket
10528505
View Documents
Thermo Scientific™
981822
uTrol Control, High
€ 229.00
Pack of 5
Estimated Shipment: 14-02-2025
to see stock.
Add to basket
Description
Description
Ensure accurate test results with Thermo Scientific™ Indiko™ and Konelab™ System Calibrators and Controls. Indiko and Konelab analyzers are fully automated for routine and special testing, including specific proteins, drugs of abuse and therapeutic drug monitoring. All Indiko™ and Konelab™ methods are based on carefully designed applications using barcoded system reagents packed for optimal efficiency.
Highlights:
  • Based on serum or urine of human or bovine origin
  • Provided in a lyophilized form
Ordering information: Not available in the U.S.A.
Specifications
Specifications
Compliant with the IVDD; CE marked
Serum, Urine