Learn More
Thermo Scientific™ Indiko™ and Konelab™ System Calibrators and Controls
Ensure accurate test results with Thermo Scientific™ Indiko™ and Konelab™ System Calibrators and Controls.
€ 229.00
Specifications
Certifications/Compliance | Compliant with the IVDD; CE marked |
---|---|
Sample Type | Serum, Urine |
Product Code | Brand | Description | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Description | Price | Quantity & Availability | |||||
10262285
|
Thermo Scientific™
981821 |
uTrol Control |
€ 229.00
Pack of 5 |
Estimated Shipment: 18-02-2025
Log in to see stock. |
|||||
10528505
|
Thermo Scientific™
981822 |
uTrol Control, High |
€ 229.00
Pack of 5 |
Estimated Shipment: 14-02-2025
Log in to see stock. |
|||||
Description
Ensure accurate test results with Thermo Scientific™ Indiko™ and Konelab™ System Calibrators and Controls. Indiko and Konelab analyzers are fully automated for routine and special testing, including specific proteins, drugs of abuse and therapeutic drug monitoring. All Indiko™ and Konelab™ methods are based on carefully designed applications using barcoded system reagents packed for optimal efficiency.
Highlights:- Based on serum or urine of human or bovine origin
- Provided in a lyophilized form
Specifications
Compliant with the IVDD; CE marked | |
Serum, Urine |