Learn More
Thermo Scientific™ Indiko™ and Konelab™ System Calibrators and Controls
Ensure accurate test results with Thermo Scientific™ Indiko™ and Konelab™ System Calibrators and Controls.
Brand:  Thermo Scientific™ 981822
 View more versions of this product
Product Code. 10528505
  • € 229.00 / Pack of 5
Estimated Shipment: 12-12-2024
to see stock.
Explore more special offers
Add to basket
Description
Description
Ensure accurate test results with Thermo Scientific™ Indiko™ and Konelab™ System Calibrators and Controls. Indiko and Konelab analyzers are fully automated for routine and special testing, including specific proteins, drugs of abuse and therapeutic drug monitoring. All Indiko™ and Konelab™ methods are based on carefully designed applications using barcoded system reagents packed for optimal efficiency.
Highlights:
  • Based on serum or urine of human or bovine origin
  • Provided in a lyophilized form
Ordering information:
  • Nortrol, Abtrol and sCal are available in the U.S.A.
  • All other calibrators and controls are not available in the U.S.A.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Compliant with the IVDD; CE marked
uTrol
Control
uTrol Control, High
Serum, Urine
10 x 5 mL
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers