Learn More
HTBA, MedChemExpress
Click to view available options
Quantity:
100 mg
500 mg
Unit Size:
100mg
500mg
Description
HTBA(3-Hydroxy-2,4,6-triiodobenzoic acid) for your research needs.
Specifications
Specifications
| Chemical Name or Material | HTBA |
| CAS | 53279-72-4 |
| Color | Off-White |
| Purity Grade Notes | Research |
| Molecular Formula | C7H3I3O3 |
| Quantity | 100 mg |
| Solubility Information | DMSO : ≥ 33 mg/mL (63.98 mM) |
| SMILES | O=C(O)C1=C(I)C=C(I)C(O)=C1I |
| Molecular Weight (g/mol) | 515.81 |
| Formula Weight | 515.81 |
| Show More |
Research purposes only