Learn More
Huenersdorff™ HDPE Universal Container
Store and transport your laboratory items in the Huenersdorf™ HDPE Universal Container. It is resistant to acids and chemicals, has a tightly closing lid, and is deep freezable and stackable.
Brand: Huenersdorff™ 911400
This item is not returnable.
View return policy
Description
- Universal container
- Natural for varied applications
- Made of food-safe polyethylene
Specifications
1000 mL | |
Hooded Lid | |
Polyethylene | |
Food Safe | |
Tightly Closing Lid | |
Container | |
110 mm |
LDPE | |
Natural | |
Round | |
Deep Freezer, Dishwasher, Microwave | |
Stackable | |
20 Pack | |
136 mm |