Learn More
Tocris Bioscience™ GSK 1838705
Potent IR and IGF1R inhibitor; also inhibits anaplastic lymphoma kinase (ALK)
Brand: Tocris Bioscience™ 5111/10
225.48 EUR valid until 2024-12-16
Use promo code "21630" to get your promotional price.
This item is not returnable.
View return policy
Description
GSK 1838705 is a potent insulin receptor (IR) and insulin-like growth factor-1 receptor (IGF1R) inhibitor (IC50 values are 1.6 and 2 nM, respectively). Also inhibits anaplastic lymphoma kinase (ALK) (IC50 = 0.5 nM). Displays > 800-fold selectivity for IR, IGFR1 and ALK over a panel of 44 kinases including JNK. Blocks proliferation of cancer cell lines in vitro, and causes complete regression of ALK-dependent tumors in vivo. Orally bioavailable.Specifications
>98% | |
GSK 1838705 | |
GSK 1838705 |
10 mg | |
532.57 |
For Research Use Only