Learn More
Poeppelmann™ GPN 680 Series Yellow LDPE Flange Cap
Brand:  Poeppelmann™ 68000250000
 View more versions of this product
Product Code. 10586825
  • € 137.00 / Pack of 100
Estimated Shipment: 19-02-2025
to see stock.
Explore more special offers
Add to basket
Specifications
Yellow
Suitable for fittings wit solid lining or tapered bore holes
19 mm
I.D. x H: 115 x 19 mm
Flange Caps
115 mm
For pipe sections with flanged end, especially valves and fittings having a solid lining (e.g., lead or Rubber) or conical bores
100 Pack
LDPE
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers