Learn More
Poeppelmann™ KAPSTO™ GPN 670 Flange Covers
Closes the opening and covers the sealing face and internal threads while preventing accidental dislodging. Poeppelmann™ KAPSTO™ GPN 670 Flange Covers are manufactured from yellow PE-LLD/PE-HD plastic.
Brand:  Poeppelmann™ 67002500000
 View more versions of this product
Product Code. 10686245
  • € 383.00 / Pack of 25
Estimated Shipment: 04-10-2024
to see stock.
Explore more special offers
Description
  • Acc. with DIN EN 12351
  • Larger tolerances accomodated by ribs
  • Suitable for PN 10 through PN 400 and DN 10 through DN 250
Specifications
Specifications
Yellow
Suitable for BASF-Werknorm 14-150. and screw holes
400 mm
Flange Cover Plug
Closes the opening and covers sealing face and internal threads
25 Pack
LDPE