Learn More
Poeppelmann™ KAPSTO™ GPN 620 Universal Protection Caps/Plugs
Featuring a tapered design for accomodating tolerances. Poeppelmann™ KAPSTO™ GPN 620 Universal Protection Caps/Plugs are manufactured from yellow PE-LD plastic.
€ 15.40 - € 55.65
Specifications
Product Type | Universal Protection Closure |
---|---|
For Use With (Application) | For a variety of applications as a plug or cap usable |
Product Code | Brand | Size (Metric) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Size (Metric) | Price | Quantity & Availability | |||||
10717435
|
Poeppelmann™
62000030000 |
11 H x 7.8 I.D. x 25 mm O.D. |
€ 15.40
Pack of 100 |
Estimated Shipment: 18-02-2025
Log in to see stock. |
|||||
10697405
|
Poeppelmann™
62000150000 |
13 H x 17.8 I.D. x 34 mm O.D. |
€ 27.95
Pack of 100 |
Estimated Shipment: 04-02-2025
Log in to see stock. |
|||||
10768565
|
Poeppelmann™
62000100000 |
12 H x 13.3 I.D. x 30 mm O.D. |
€ 18.85
Pack of 100 |
Estimated Shipment: 04-02-2025
Log in to see stock. |
|||||
10093714
|
Poeppelmann™
62000170000 |
15 H x 19.5 I.D. x 37 mm O.D. |
€ 20.50
Pack of 100 |
Estimated Shipment: 04-02-2025
Log in to see stock. |
|||||
10013524
|
Poeppelmann™
62000070000 |
10 H x 10.6 I.D. x 28 mm O.D. |
€ 16.85
Pack of 100 |
Estimated Shipment: 25-02-2025
Log in to see stock. |
|||||
10308584
|
Poeppelmann™
62000180000 |
15 H x 20.1 I.D. x 37 mm O.D. |
€ 27.00
Pack of 100 |
Estimated Shipment: 17-02-2025
Log in to see stock. |
|||||
10431785
|
Poeppelmann™
62000230000 |
19 H x 26.4 I.D. x 44 mm O.D. |
€ 48.90
Pack of 100 |
Estimated Shipment: 23-01-2025
Log in to see stock. |
|||||
10638175
|
Poeppelmann™
62000250000 |
19 H x 29.7 I.D. x 47 mm O.D. |
€ 55.65
Pack of 100 |
Estimated Shipment: 03-02-2025
Log in to see stock. |
|||||
10105134
|
Poeppelmann™
62000260000 |
19 H x 31.2 I.D. x 49 mm O.D. |
€ 48.15
Pack of 100 |
Estimated Shipment: 25-02-2025
Log in to see stock. |
|||||
Description
- Can be used as plug or cap
- For a wide range of applications
- Produced in acc. with VDA Band 19 and ISO 16232
- Same design as GPN 610 but with a larger cover flange
Specifications
Universal Protection Closure | |
For a variety of applications as a plug or cap usable |