Learn More
Poeppelmann™ KAPSTO™ GPN 620 Universal Protection Caps/Plugs
Featuring a tapered design for accomodating tolerances. Poeppelmann™ KAPSTO™ GPN 620 Universal Protection Caps/Plugs are manufactured from yellow PE-LD plastic.
Brand:  Poeppelmann™ 62000100000
 View more versions of this product
Product Code. 10768565
  • € 18.85 / Pack of 100
Estimated Shipment: 04-02-2025
to see stock.
Explore more special offers
Description
  • Can be used as plug or cap
  • For a wide range of applications
  • Produced in acc. with VDA Band 19 and ISO 16232
  • Same design as GPN 610 but with a larger cover flange
Specifications
Specifications
Universal Protection Closure
For a variety of applications as a plug or cap usable
12 H x 13.3 I.D. x 30 mm O.D.