Learn More
Poeppelmann™ KAPSTO™ GPN 600 Tapered Closures
Excellent for closing off bore holes and internal threads, or to protect studs and external threads. Poeppelmann™ KAPSTO™ GPN 600 Tapered Closures are manufactured from natural-colored PE-LD plastic.
Brand: Poeppelmann™ 60029900000
Description
- Universal use as plug or cap
- Tapered design ideal for accomodating tolerances
- Chemically resistant to alcohol and weak acids/alkalis
Specifications
Natural | |
22.5 mm | |
O.D. x H: 315 x 22.5 mm | |
LDPE |
For use as plugs in bores or internal thread, or as caps for external thread | |
25 Pack | |
315 mm | |
Tapered Closure |