Learn More
Poeppelmann™ KAPSTO™ GPN 600 Tapered Closures
Excellent for closing off bore holes and internal threads, or to protect studs and external threads. Poeppelmann™ KAPSTO™ GPN 600 Tapered Closures are manufactured from natural-colored PE-LD plastic.
Brand:  Poeppelmann™ 60007400000
 View more versions of this product
Product Code. 10195314
  • € 184.37 / Pack of 100
In Stock
to see stock.
Edge
Explore more special offers
Add to basket
Description
Description
  • Universal use as plug or cap
  • Tapered design ideal for accomodating tolerances
  • Chemically resistant to alcohol and weak acids/alkalis
Specifications
Specifications
Natural
17 mm
O.D. x H: 85 x 17 mm
LDPE
For use as plugs in bores or internal thread, or as caps for external thread
100 Pack
85 mm
Tapered Closure
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Poeppelmann™ KAPSTO™ GPN 600 Tapered Closures > Size (O.D. x H): 85 x 17mm