Learn More
Poeppelmann™ GPN 600 Series Natural LDPE Tapered Closure (7mm Insertion Depth)
Brand:  Poeppelmann™ 60001330000
 View more versions of this product
Product Code. 10359284
  • € 11.65 / Pack of 100
In Stock
to see stock.
Edge
Explore more special offers
Specifications
Natural
6.8 mm
O.D. x H: 16 x 6.8 mm
LDPE
For use as plugs in bores or internal thread, or as caps for external thread
100 Pack
16 mm
Tapered Closure