Learn More
Poeppelmann™ GPN 600 Series Natural LDPE Tapered Closure (7mm Insertion Depth)
Brand:  Poeppelmann™ 60001330000
 View more versions of this product
Product Code. 10359284
  • € 11.65 / Pack of 100
In Stock
to see stock.
Edge
Explore more special offers
Add to basket
Specifications
Natural
6.8 mm
O.D. x H: 16 x 6.8 mm
LDPE
For use as plugs in bores or internal thread, or as caps for external thread
100 Pack
16 mm
Tapered Closure
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers