Learn More
VEE GEE Glass Thermometer
The serialized glass thermometer is a precise instrument suitable for various general-purpose applications in laboratories, universities, food and beverage, environmental, wastewater, and other settings
€ 34.85 - € 43.30
Specifications
Bottle Fill | Safety Blue |
---|---|
For Use With (Application) | Laboratories, universities, food/beverage, environmental, wastewater and many other general-purpose applications |
Material | Glass |
Probe Type | Immersion/General Purpose (Tapered) |
Thermometer Fill | Safety Blue |
Product Code | Brand | Immersion Type | Temperature Range (English) | Temperature Range (Metric) | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Immersion Type | Temperature Range (English) | Temperature Range (Metric) | Price | Quantity & Availability | |||
17821224
|
VEE GEE
80701 |
Total Immersion | -30 to +120°F | -34.4 to +48.9°C |
€ 34.85
Each |
Estimated Shipment: 08-01-2025
Log in to see stock. |
|||
17811224
|
VEE GEE
80702 |
Total Immersion | -4 to +230°F | -20 to +110°C |
€ 34.85
Each |
Estimated Shipment: 08-01-2025
Log in to see stock. |
|||
17831224
|
VEE GEE
80703 |
Total Immersion | 0 to 300°F | -17.8 to +148.9°C |
€ 34.85
Each |
Estimated Shipment: 08-01-2025
Log in to see stock. |
|||
17801224
|
VEE GEE
80706 |
Total Immersion | -31 to +122°F | -35 to +50°C |
€ 34.85
Each |
Estimated Shipment: 08-01-2025
Log in to see stock. |
|||
17891214
|
VEE GEE
80707 |
Total Immersion | -4 to +230°F | -20 to +110°C |
€ 34.85
Each |
Estimated Shipment: 08-01-2025
Log in to see stock. |
|||
17831194
|
VEE GEE
80708 |
Total Immersion | -4 to +302°F | -20 to +150°C |
€ 34.85
Each |
Estimated Shipment: 08-01-2025
Log in to see stock. |
|||
17891254
|
VEE GEE
80901 |
Partial Immersion | -30 to +120°F | -34.4 to +48.9°C |
€ 34.85
Each |
Estimated Shipment: 08-01-2025
Log in to see stock. |
|||
17891224
|
VEE GEE
80902 |
Partial Immersion | -4 to +230°F | -20 to +110°C |
€ 34.85
Each |
Estimated Shipment: 08-01-2025
Log in to see stock. |
|||
17881224
|
VEE GEE
80903 |
Partial Immersion | 0 to 300°F | -17.8 to +148.9°C |
€ 34.85
Each |
Estimated Shipment: 08-01-2025
Log in to see stock. |
|||
17841224
|
VEE GEE
80905 |
Partial Immersion | 20 to 500°F | -6.7 to +260°C |
€ 43.30
Each |
Estimated Shipment: 08-01-2025
Log in to see stock. |
|||
17871224
|
VEE GEE
80906 |
Partial Immersion | -31 to +122°F | -35 to +50°C |
€ 34.85
Each |
Estimated Shipment: 08-01-2025
Log in to see stock. |
|||
17851224
|
VEE GEE
80907 |
Partial Immersion | -4 to +230°F | -20 to +110°C |
€ 34.85
Each |
Estimated Shipment: 08-01-2025
Log in to see stock. |
|||
17821254
|
VEE GEE
80908 |
Partial Immersion | -4 to +302°F | -20 to +150°C |
€ 34.85
Each |
Estimated Shipment: 08-01-2025
Log in to see stock. |
|||
17851234
|
VEE GEE
80910 |
Partial Immersion | -10 to +250°F | -23.2 to +121°C |
€ 43.30
Each |
Estimated Shipment: 08-01-2025
Log in to see stock. |
|||
Description
- Feature Safety Blue fill against a white background for easy reading
- Accuracy is equal to one subdivision
- Immersion: Total or Partial option
Specifications
Safety Blue | |
Glass | |
Safety Blue | |
0.64 cm |
Laboratories, universities, food/beverage, environmental, wastewater and many other general-purpose applications | |
Immersion/General Purpose (Tapered) | |
±1 subdivision |