Learn More
DWK Life Sciences Kimble™ Ultra-Ware™ Glass Support Base
The Kimble™ Ultra-Ware™ Glass Support Base is suitable for use with Kimble™ Ultra-Ware™ Microfiltration Assemblies.
Brand:  DWK Life Sciences 953726-0001
Product Code. 11808502
  • € 115.00 / Each
Estimated Shipment: 10-01-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Product Type: Support Base
  • Material: Glass
  • Size: 25 mm
  • For Use with: Kimble™ Ultra-Ware™ Microfiltration Assembly
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Not Autoclavable
Kimble Ultra-Ware Microfiltration Assembly
Non-sterile
25 mm
Reusable
Glass
Support Base
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers