Learn More
BUCHI Glass Assembly A
Brand:  BUCHI 048169
Product Code. 12726854
  • € 1095.00 / Each
Explore more special offers
Specifications
Glass
For R-210 and R-215 Rotavapors
Plastic and Glass
1 L
Glass Assembly A
Receiving flask
1 Pack
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
BUCHI Glass Assembly A > Capacity: 1L