Learn More
Ginkgolide C, MedChemExpress
MedChemExpress Ginkgolide C is a flavone isolated from Ginkgo biloba leaves, possessing multiple biological functions, such as decreasing platelet aggregation and ameliorating Alzheimer disease.
€ 169.50 - € 468.00
Specifications
Chemical Name or Material | Ginkgolide C |
---|---|
CAS | 15291-76-6 |
Color | White |
Purity Grade Notes | Research |
Molecular Formula | C20H24O11 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16494887
|
Medchem Express
HY-N0785 |
50 mg |
€ 468.00
50mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16479098
|
Medchem Express
HY-N0785 |
10 mg |
€ 169.50
10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Ginkgolide C is a flavone isolated from Ginkgo biloba leaves, possessing multiple biological functions, such as decreasing platelet aggregation and ameliorating Alzheimer disease.Specifications
Ginkgolide C | |
White | |
C20H24O11 | |
DMSO : 250 mg/mL (567.67 mM; Need ultrasonic) | |
440.4 | |
98.0% | |
Solid |
15291-76-6 | |
Research | |
BN-52022 Ginkgolide-C | |
O[C@@H]1C2([C@@](O3)([H])[C@H](O)[C@H]4C(C)(C)C)C4([C@H]5O)[C@](OC5=O)([H])OC2(C3=O)[C@@](O)([C@@H]6C)[C@@]1([H])OC6=O | |
440.4 | |
Research | |
Metabolism-sugar/lipid metabolism |
Safety and Handling
H302∣H315∣H319∣H335
missing translation for 'shelfLife' : Powder -20°C 3 years, 4°C 2 years∣In solvent -80°C 6 months, -20°C 1 month
Recommended Storage : Powder -20°C 3 years, 4°C 2 years∣In solvent -80°C 6 months, -20°C 1 month
Research purposes only
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.