Learn More
Ginkgolide C, MedChemExpress
MedChemExpress Ginkgolide C is a flavone isolated from Ginkgo biloba leaves, possessing multiple biological functions, such as decreasing platelet aggregation and ameliorating Alzheimer disease.
Brand: Medchem Express HY-N0785
This item is not returnable.
View return policy
Description
Ginkgolide C is a flavone isolated from Ginkgo biloba leaves, possessing multiple biological functions, such as decreasing platelet aggregation and ameliorating Alzheimer disease.Specifications
Ginkgolide C | |
White | |
C20H24O11 | |
BN-52022 Ginkgolide-C | |
O[C@@H]1C2([C@@](O3)([H])[C@H](O)[C@H]4C(C)(C)C)C4([C@H]5O)[C@](OC5=O)([H])OC2(C3=O)[C@@](O)([C@@H]6C)[C@@]1([H])OC6=O | |
440.4 | |
Research | |
Metabolism-sugar/lipid metabolism |
15291-76-6 | |
Research | |
10 mg | |
DMSO : 250 mg/mL (567.67 mM; Need ultrasonic) | |
440.4 | |
98.0% | |
Solid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Research purposes only