Learn More
Ginkgolide C, MedChemExpress
MedChemExpress Ginkgolide C is a flavone isolated from Ginkgo biloba leaves, possessing multiple biological functions, such as decreasing platelet aggregation and ameliorating Alzheimer disease.
Brand:  Medchem Express HY-N0785
Encompass
 View more versions of this product
Product Code. 16479098
  • € 169.50 / 10mg
Explore more special offers
Description
Description
Ginkgolide C is a flavone isolated from Ginkgo biloba leaves, possessing multiple biological functions, such as decreasing platelet aggregation and ameliorating Alzheimer disease.
Specifications
Specifications
Ginkgolide C
White
C20H24O11
BN-52022 Ginkgolide-C
O[C@@H]1C2([C@@](O3)([H])[C@H](O)[C@H]4C(C)(C)C)C4([C@H]5O)[C@](OC5=O)([H])OC2(C3=O)[C@@](O)([C@@H]6C)[C@@]1([H])OC6=O
440.4
Research
Metabolism-sugar/lipid metabolism
15291-76-6
Research
10 mg
DMSO : 250 mg/mL (567.67 mM; Need ultrasonic)
440.4
98.0%
Solid
Certificates
Special Offers
Special Offers
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Research purposes only